Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHACTR1 SIRNA Assays ›
Gene Symbol:
PHACTR1
Gene Name:
phosphatase and actin regulator 1
Gene Aliases:
RPEL, RPEL1, dJ257A7.2
Chromosome Location:
Chr.6: 12716635 - 13290484 on Build GRCh38
UniGene ID:
Hs.436996
Species:
Human
siRNA ID
HSS176836
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHACTR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242648.2
NP_001229577.1
14
2001
NM_001322308.1
NP_001309237.1
13
1956
NM_001322309.1
NP_001309238.1
13
1847
NM_001322310.1
NP_001309239.1
13
1922
NM_001322311.1
NP_001309240.1
11
1608
NM_001322312.1
NP_001309241.1
11
1971
NM_001322313.1
NP_001309242.1
12
2178
NM_001322314.1
NP_001309243.1
12
2110
NM_030948.3
NP_112210.1
14
2191
XM_005248934.2
XP_005248991.1
15
2240
XM_017010452.1
XP_016865941.1
12
1904
XM_017010454.1
XP_016865943.1
15
2240
XM_017010455.1
XP_016865944.1
15
2208
XM_017010456.1
XP_016865945.1
14
2205
XM_017010457.1
XP_016865946.1
14
2184
XM_017010458.1
XP_016865947.1
11
1697
XM_017010459.1
XP_016865948.1
14
2033
XM_017010460.1
XP_016865949.1
14
2001
XM_017010461.1
XP_016865950.1
13
1972
XM_017010462.1
XP_016865951.1
13
1998
XM_017010463.1
XP_016865952.1
12
2098
XM_017010464.1
XP_016865953.1
11
1697
XM_017010465.1
XP_016865954.1
12
1878
XM_017010466.1
XP_016865955.1
12
1891
XM_017010467.1
XP_016865956.1
11
1878
XM_017010469.1
XP_016865958.1
11
1680
GenBank mRNA
AK090769.1
-
13
1858
AK290017.1
-
14
1852
BC101526.1
-
13
1765
8069032
BC101528.1
-
13
1765
8069034
BF026507.1
-
372
BQ434462.1
-
4
408
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
actomyosin structure organization
actin cytoskeleton reorganization
negative regulation of catalytic activity
stress fiber assembly
cell motility
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0