Gene Symbol: PXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183373.3 | NP_899229.2 | 5 | 1126 | ||
XM_011514393.2 | XP_011512695.1 | 7 | 1105 | |||
GenBank mRNA | AJ420534.1 | - | 1 | 532 | ||
AK024828.1 | - | 5 | 599 | |||
AK289598.1 | - | 5 | 827 | |||
AL832792.1 | - | 5 | 672 | |||
BC045715.2 | - | 4 | 390 | |||
BC056908.1 | - | 5 | 816 | 6158428 |