Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LHFPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182548.3 | NP_872354.1 | 2 | 805 | ||
GenBank mRNA | AK126841.1 | - | 2 | 720 | ||
BC028630.2 | - | 2 | 805 | 5262164 |