Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014904.2 | NP_055719.1 | 3 | 1442 | ||
XM_005269629.3 | XP_005269686.1 | 3 | 1760 | |||
XM_017015926.1 | XP_016871415.1 | 3 | 1760 | |||
GenBank mRNA | AB023158.1 | - | 3 | 1442 | ||
AY037299.1 | - | 3 | 1442 | |||
BC075073.2 | - | 3 | 1044 | 30915593 | ||
BC075074.2 | - | 3 | 1044 | 30915380 | ||
DQ013303.1 | - | 3 | 1002 |