Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033053.2 | NP_001028225.1 | 13 | 4167 | ||
NM_014922.4 | NP_055737.1 | 13 | 4155 | |||
NM_033004.3 | NP_127497.1 | 13 | 4155 | |||
NM_033006.3 | NP_127499.1 | 12 | 4065 | |||
NM_033007.3 | NP_127500.1 | 12 | 4065 | |||
GenBank mRNA | AB023143.1 | - | 13 | 4122 | ||
AF229059.1 | - | 13 | 3600 | |||
AF229060.1 | - | 13 | 3600 | |||
AF229061.1 | - | 12 | 3510 | |||
AF229062.1 | - | 12 | 3510 | |||
AF298548.1 | - | 14 | 3759 | |||
AF310105.1 | - | 13 | 4118 | |||
AK026393.1 | - | 10 | 2078 | |||
AK026398.1 | - | 10 | 1428 | |||
AL117470.1 | - | 10 | 2758 |