Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014939.3 | NP_055754.2 | 9 | 1694 | ||
XM_005258233.2 | XP_005258290.1 | 9 | 1787 | |||
XM_005258234.2 | XP_005258291.1 | 9 | 1787 | |||
XM_005258235.3 | XP_005258292.1 | 4 | 568 | |||
XM_006722420.3 | XP_006722483.1 | 9 | 1787 | |||
XM_011525884.2 | XP_011524186.1 | 9 | 1787 | |||
XM_017025615.1 | XP_016881104.1 | 9 | 1286 | |||
XM_017025616.1 | XP_016881105.1 | 9 | 1784 | |||
XM_017025617.1 | XP_016881106.1 | 9 | 1784 | |||
GenBank mRNA | AB023229.1 | - | 9 | 1387 | ||
AK001662.1 | - | 1 | 53 | |||
AK292537.1 | - | 9 | 1673 | |||
AL136749.1 | - | 10 | 1481 | |||
BC059380.1 | - | 9 | 1673 | |||
BC127109.1 | - | 9 | 1248 | 40124744 | ||
BC127110.1 | - | 9 | 1248 | 40124745 |