Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: FKBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002014.3 | NP_002005.1 | 4, 5 | 719 | ||
XM_011520929.2 | XP_011519231.1 | 4, 5 | 1712 | |||
GenBank mRNA | AK313918.1 | - | 4, 5 | 684 | ||
BC001786.1 | - | 4, 5 | 674 | 3542330 | ||
BC002887.2 | - | 1, 2 | 78 | |||
BC007924.2 | - | 4, 5 | 619 | 4304462 | ||
M88279.1 | - | 4, 5 | 620 |