Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014951.2 | NP_055766.2 | 2 | 425 | ||
NM_199450.2 | NP_955522.1 | 2 | 425 | |||
NM_199451.2 | NP_955523.1 | 2 | 425 | |||
XM_017015937.1 | XP_016871426.1 | 2 | 973 | |||
GenBank mRNA | AB020651.1 | - | 2 | 387 | ||
AJ505147.1 | - | 2 | 198 | |||
AJ505148.1 | - | 2 | 387 | |||
AJ505149.1 | - | 2 | 387 | |||
AK289515.1 | - | 2 | 166 | |||
AK289808.1 | - | 2 | 190 | |||
BC017841.1 | - | 2 | 229 | |||
BC070073.1 | - | 2 | 210 | |||
BX647853.1 | - | 2 | 218 | |||
BX647904.1 | - | 2 | 218 | |||
DB476702.1 | - | 2 | 422 |