Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP164 SIRNA Assays ›
Gene Symbol:
CEP164
Gene Name:
centrosomal protein 164
Gene Aliases:
NPHP15
Chromosome Location:
Chr.11: 117316346 - 117413266 on Build GRCh38
UniGene ID:
Hs.504009
Species:
Human
siRNA ID
HSS177012
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP164
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271933.1
NP_001258862.1
23, 24
3187
NM_014956.4
NP_055771.4
24, 25
3244
XM_005271453.1
XP_005271510.1
25, 26
5201
XM_005271456.1
XP_005271513.1
24, 25
3515
XM_005271457.1
XP_005271514.1
24, 25
3524
XM_006718788.1
XP_006718851.1
25, 26
5210
XM_006718794.1
XP_006718857.1
23, 24
3437
XM_011542674.1
XP_011540976.1
25, 26
5261
XM_017017364.1
XP_016872853.1
25, 26
5128
XM_017017365.1
XP_016872854.1
25, 26
5280
XM_017017366.1
XP_016872855.1
25, 26
5063
XM_017017367.1
XP_016872856.1
24, 25
5189
XM_017017369.1
XP_016872858.1
25, 26
5261
XM_017017371.1
XP_016872860.1
24, 25
5183
XM_017017372.1
XP_016872861.1
24, 25
5123
XM_017017373.1
XP_016872862.1
24, 25
5061
XM_017017374.1
XP_016872863.1
24, 25
4856
XM_017017375.1
XP_016872864.1
23, 24
4985
XM_017017376.1
XP_016872865.1
24, 25
5114
XM_017017377.1
XP_016872866.1
26, 27
5220
XM_017017378.1
XP_016872867.1
23, 24
4904
XM_017017379.1
XP_016872868.1
24, 25
3575
XM_017017380.1
XP_016872869.1
23, 24
3237
XM_017017381.1
XP_016872870.1
23, 24
3497
XM_017017382.1
XP_016872871.1
22, 23
3159
XM_017017383.1
XP_016872872.1
23, 24
3426
XM_017017384.1
XP_016872873.1
23, 24
3225
XM_017017385.1
XP_016872874.1
21, 22
2981
XM_017017386.1
XP_016872875.1
21, 22
3141
GenBank mRNA
AB028975.1
-
23, 24
3458
AK125995.1
-
18, 19
3332
AK302991.1
-
6, 7
1180
AK303067.1
-
22, 23
3098
AL117632.1
-
5, 6
506
AL137265.1
-
17, 18
2421
BX647583.1
-
10, 11
3979
BX648682.1
-
24, 25
3244
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
DNA repair
mitotic nuclear division
cilium assembly
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0