Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318226.1 | NP_001305155.1 | 15, 16 | 2007 | ||
NM_025176.5 | NP_079452.3 | 15, 16 | 2007 | |||
XM_011529186.2 | XP_011527488.1 | 15, 16 | 2645 | |||
XM_011529187.2 | XP_011527489.1 | 15, 16 | 2630 | |||
XM_011529188.1 | XP_011527490.1 | 15, 16 | 2022 | |||
XM_011529189.1 | XP_011527491.1 | 15, 16 | 2030 | |||
XM_011529190.1 | XP_011527492.1 | 14, 15 | 1945 | |||
XM_011529191.1 | XP_011527493.1 | 11, 12 | 1537 | |||
XM_011529192.1 | XP_011527494.1 | 8, 9 | 2457 | |||
XM_011529193.1 | XP_011527495.1 | 14, 15 | 1945 | |||
XM_011529194.2 | XP_011527496.1 | 6, 7 | 1726 | |||
XM_011529195.2 | XP_011527497.1 | 5, 6 | 551 | |||
XM_011529197.2 | XP_011527499.1 | 7, 8 | 920 | |||
XM_017027732.1 | XP_016883221.1 | 16, 17 | 3964 | |||
XM_017027733.1 | XP_016883222.1 | 16, 17 | 2076 | |||
XM_017027734.1 | XP_016883223.1 | 9, 10 | 1111 | |||
XM_017027735.1 | XP_016883224.1 | 5, 6 | 556 | |||
GenBank mRNA | AB023197.1 | - | 15, 16 | 1993 | ||
AK093569.1 | - | 5, 6 | 1390 | |||
AK309298.1 | - | 15, 16 | 1956 | |||
BC036380.2 | - | 14, 15 | 1945 | 5268454 | ||
EU718622.1 | - | 15, 16 | 1993 |