Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015002.2 | NP_055817.1 | 5, 6 | 781 | ||
NM_033624.2 | NP_296373.1 | 5, 6 | 781 | |||
XM_017019037.1 | XP_016874526.1 | 5, 6 | 786 | |||
XM_017019038.1 | XP_016874527.1 | 5, 6 | 786 | |||
XM_017019039.1 | XP_016874528.1 | 4, 5 | 397 | |||
GenBank mRNA | AB020682.1 | - | 5, 6 | 743 | ||
AF174601.1 | - | 5, 6 | 742 | |||
AK001699.1 | - | 5, 6 | 765 | |||
AK074714.1 | - | 2, 3 | 250 | |||
AK308141.1 | - | 6, 7 | 688 | |||
AK314573.1 | - | 5, 6 | 754 | |||
AL136899.1 | - | 5, 6 | 594 | |||
BC034045.1 | - | 6, 7 | 708 | 4815947 | ||
BC042333.1 | - | 6, 7 | 681 | 5195515 | ||
BC091496.1 | - | 5, 6 | 740 |