Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009999.2 | NP_001009999.1 | 5 | 908 | ||
NM_015013.3 | NP_055828.2 | 4 | 848 | |||
XM_005245786.2 | XP_005245843.1 | 5 | 913 | |||
XM_006710472.3 | XP_006710535.1 | 4 | 853 | |||
XM_006710473.2 | XP_006710536.1 | 5 | 913 | |||
XM_006710474.3 | XP_006710537.1 | 5 | 913 | |||
XM_017000716.1 | XP_016856205.1 | 5 | 913 | |||
XM_017000717.1 | XP_016856206.1 | 4 | 853 | |||
GenBank mRNA | AB011173.1 | - | 4 | 801 | ||
AK290328.1 | - | 4 | 374 | |||
BC016639.1 | - | 2 | 87 | |||
BC040194.2 | - | 5 | 863 | 4815528 | ||
BC048134.1 | - | 4 | 769 | 5298150 |