Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015017.4 | NP_055832.3 | 18, 19 | 2384 | ||
NM_201624.2 | NP_963918.1 | 17, 18 | 2205 | |||
NM_201626.2 | NP_963920.1 | 18, 19 | 2360 | |||
XM_005270648.1 | XP_005270705.1 | 17, 18 | 2008 | |||
XM_005270649.1 | XP_005270706.1 | 17, 18 | 2032 | |||
XM_017000722.1 | XP_016856211.1 | 17, 18 | 2181 | |||
XM_017000723.1 | XP_016856212.1 | 17, 18 | 2156 | |||
XM_017000724.1 | XP_016856213.1 | 17, 18 | 2181 | |||
XM_017000727.1 | XP_016856216.1 | 18, 19 | 2100 | |||
XM_017000728.1 | XP_016856217.1 | 18, 19 | 2062 | |||
GenBank mRNA | AB029020.1 | - | 17, 18 | 2126 | ||
AF383172.1 | - | 18, 19 | 2357 | |||
AF383173.1 | - | 17, 18 | 2178 | |||
AK022864.1 | - | 18, 19 | 2333 | |||
AK023664.1 | - | 8, 9 | 824 | |||
AK225499.1 | - | 15, 16 | 1825 | |||
AL050140.1 | - | 12 | ||||
BC016663.1 | - | 18, 19 | 2214 | 3874822 | ||
BU631866.1 | - | 4, 5 | 596 |