Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015589.5 | NP_056404.4 | 3 | 1217 | ||
GenBank mRNA | AK024652.1 | - | 3 | 910 |