Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015055.3 | NP_055870.2 | 3, 4 | 520 | ||
GenBank mRNA | AB014540.1 | - | 3, 4 | 472 | ||
AF134894.1 | - | 3, 4 | 491 | |||
AF210818.1 | - | 3, 4 | 417 | |||
AK304104.1 | - | 3, 4 | 384 | |||
BC000616.2 | - | 3, 4 | 493 | 3346240 | ||
DA413747.1 | - | 3, 4 | 520 |