Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L3 SIRNA Assays ›
Gene Symbol:
EPB41L3
Gene Name:
erythrocyte membrane protein band 4.1 like 3
Gene Aliases:
4.1B, DAL-1, DAL1
Chromosome Location:
Chr.18: 5392381 - 5630666 on Build GRCh38
UniGene ID:
Hs.213394
Species:
Human
siRNA ID
HSS177201
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281533.1
NP_001268462.1
16
2111
NM_012307.3
NP_036439.2
17
2799
XM_011525623.2
XP_011523925.1
17
2367
XM_011525626.2
XP_011523928.1
15
1933
XM_011525628.2
XP_011523930.1
16
2292
XM_017025618.1
XP_016881107.1
15
1933
XM_017025619.1
XP_016881108.1
15
1933
XM_017025621.1
XP_016881110.1
13
1804
XM_017025622.1
XP_016881111.1
17
2313
XM_017025623.1
XP_016881112.1
14
1858
XM_017025625.1
XP_016881114.1
16
2060
XM_017025626.1
XP_016881115.1
14
1858
XM_017025627.1
XP_016881116.1
17
2367
XM_017025628.1
XP_016881117.1
15
2256
XM_017025629.1
XP_016881118.1
16
2238
XM_017025630.1
XP_016881119.1
16
2006
XM_017025631.1
XP_016881120.1
13
1804
XM_017025632.1
XP_016881121.1
16
2292
XM_017025633.1
XP_016881122.1
16
2060
XM_017025636.1
XP_016881125.1
16
2292
XM_017025639.1
XP_016881128.1
16
2238
XM_017025642.1
XP_016881131.1
16
2238
XM_017025645.1
XP_016881134.1
17
2226
XM_017025647.1
XP_016881136.1
16
2190
XM_017025648.1
XP_016881137.1
16
2151
XM_017025649.1
XP_016881138.1
17
2172
XM_017025650.1
XP_016881139.1
17
2226
XM_017025651.1
XP_016881140.1
15
2115
XM_017025652.1
XP_016881141.1
16
2097
XM_017025653.1
XP_016881142.1
17
2172
XM_017025654.1
XP_016881143.1
16
2151
XM_017025655.1
XP_016881144.1
15
2061
XM_017025656.1
XP_016881145.1
16
2151
XM_017025658.1
XP_016881147.1
16
2097
GenBank mRNA
AB023204.1
-
17
2476
AK294610.1
-
5
384
AK294690.1
-
5
456
AK295453.1
-
6
477
AK297406.1
-
16
2111
AK303679.1
-
6
574
AK307751.1
-
6
566
AL832598.2
-
1
1464
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
protein localization to paranode region of axon
apoptotic process
cytoskeletal anchoring at plasma membrane
biological_process
regulation of cell shape
cortical cytoskeleton organization
cortical actin cytoskeleton organization
paranodal junction assembly
myelin maintenance
neuron projection morphogenesis
protein localization to juxtaparanode region of axon
protein localization to plasma membrane
actin binding
structural constituent of cytoskeleton
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0