Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZZEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015113.3 | NP_055928.3 | 29 | 4484 | ||
XM_005256561.2 | XP_005256618.1 | 29 | 4548 | |||
XM_011523759.2 | XP_011522061.1 | 29 | 4548 | |||
XM_017024382.1 | XP_016879871.1 | 29 | 4548 | |||
XM_017024383.1 | XP_016879872.1 | 29 | 4545 | |||
XM_017024384.1 | XP_016879873.1 | 4 | 359 | |||
GenBank mRNA | AB007859.4 | - | 29 | 4484 | ||
AK122719.1 | - | 4 | 578 | |||
BC151836.1 | - | 29 | 4484 | 8860131 |