Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
siRNA ID
HSS177350
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
131
24117
NM_182961.3
NP_892006.3
132
24495
XM_006715407.1
XP_006715470.1
132
24408
XM_006715408.2
XP_006715471.1
132
24400
XM_006715409.1
XP_006715472.1
131
24387
XM_006715410.2
XP_006715473.1
132
24412
XM_006715411.1
XP_006715474.1
131
24357
XM_006715412.2
XP_006715475.1
132
24397
XM_006715413.2
XP_006715476.1
132
24412
XM_006715414.1
XP_006715477.1
130
24336
XM_006715415.2
XP_006715478.1
132
24412
XM_006715416.2
XP_006715479.1
132
24397
XM_006715417.2
XP_006715480.1
131
24271
XM_006715420.2
XP_006715483.1
131
24259
XM_006715421.2
XP_006715484.1
131
24256
XM_006715422.1
XP_006715485.1
131
24249
XM_006715423.2
XP_006715486.1
132
24412
XM_006715424.2
XP_006715487.1
132
24412
XM_006715425.2
XP_006715488.1
132
24412
XM_011535641.2
XP_011533943.1
132
24409
XM_011535642.2
XP_011533944.1
132
24397
XM_011535643.1
XP_011533945.1
131
24243
XM_011535644.1
XP_011533946.1
118
22459
XM_011535645.2
XP_011533947.1
114
21852
XM_017010608.1
XP_016866097.1
133
24527
XM_017010609.1
XP_016866098.1
132
24334
XM_017010610.1
XP_016866099.1
131
24313
XM_017010611.1
XP_016866100.1
131
24116
XM_017010612.1
XP_016866101.1
130
24040
XM_017010613.1
XP_016866102.1
132
24409
XM_017010614.1
XP_016866103.1
131
24256
XM_017010615.1
XP_016866104.1
131
24256
XM_017010616.1
XP_016866105.1
132
24412
XM_017010617.1
XP_016866106.1
132
24409
XM_017010618.1
XP_016866107.1
132
24397
XM_017010619.1
XP_016866108.1
118
22491
GenBank mRNA
AB018339.2
-
20
3132
AF444779.1
-
7
1005
AF495910.1
-
132
24495
AF535142.1
-
131
24117
AK122691.1
-
12
1775
AK127076.1
-
16
2472
AK304825.1
-
6
697
AK316101.1
-
5
712
AY061755.1
-
47
7585
AY061756.1
-
4
575
AY184203.1
-
3
440
AY184206.1
-
45
7480
BC090927.1
-
5
757
BX537837.1
-
14
2073
BX647837.1
-
17
2667
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-hjwgp:80/10.239.166.163:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0