Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015329.3 | NP_056144.3 | 17, 18 | 1674 | ||
XM_005259837.3 | XP_005259894.2 | 17, 18 | 1663 | |||
XM_005259838.3 | XP_005259895.3 | 15, 16 | 1725 | |||
XM_011527844.2 | XP_011526146.1 | 17, 18 | 2079 | |||
XM_017026539.1 | XP_016882028.1 | 15, 16 | 1721 | |||
GenBank mRNA | AB020699.1 | - | 17, 18 | 1660 | ||
AK001349.1 | - | 5, 6 | 391 | |||
AK025896.1 | - | 15, 16 | 1326 | |||
AK125914.1 | - | 2, 3 | 1893 | |||
AK126227.1 | - | 4, 5 | 1566 | |||
AK128029.1 | - | 8, 9 | 1658 | |||
AL110250.1 | - | 6, 7 | 449 | |||
BC010222.1 | - | 6, 7 | 460 | |||
BC063863.1 | - | 6, 7 | 807 |