Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 17 | 2047 | ||
XM_005268750.4 | XP_005268807.1 | 17 | 1846 | |||
XM_011538083.1 | XP_011536385.1 | 17 | 1911 | |||
XM_017019091.1 | XP_016874580.1 | 18 | 1997 | |||
XM_017019092.1 | XP_016874581.1 | 17 | 1860 | |||
XM_017019093.1 | XP_016874582.1 | 17 | 1809 | |||
XM_017019094.1 | XP_016874583.1 | 16 | 1727 | |||
XM_017019095.1 | XP_016874584.1 | 13 | 1439 | |||
GenBank mRNA | AB023163.1 | - | 17 | 1991 | ||
AB024494.1 | - | 17 | 2537 | |||
AB097013.1 | - | 17 | 1867 | |||
AB097035.1 | - | 17 | 1867 | |||
AK292866.1 | - | 17 | 1863 | |||
BC050324.1 | - | 17 | 2006 | 5266097 |