Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314030.1 | NP_001300959.1 | 14 | 2890 | ||
NM_012284.2 | NP_036416.1 | 14 | 2894 | |||
XM_011538085.2 | XP_011536387.1 | 14 | 3543 | |||
XM_011538086.2 | XP_011536388.1 | 14 | 3539 | |||
XM_017019096.1 | XP_016874585.1 | 13 | 3309 | |||
XM_017019097.1 | XP_016874586.1 | 14 | 2514 | |||
GenBank mRNA | AB022696.1 | - | 14 | 2669 | ||
AB033108.1 | - | 14 | 2894 | |||
BC033141.1 | - | 3 | 439 | |||
BC150602.1 | - | 14 | 2775 | 9056972 |