Gene Symbol: PHF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290259.1 | NP_001277188.1 | 10 | 3616 | ||
NM_015153.3 | NP_055968.1 | 8 | 3075 | |||
XM_005248701.3 | XP_005248758.1 | 9 | 3410 | |||
XM_005248702.3 | XP_005248759.1 | 8 | 3154 | |||
XM_005248703.3 | XP_005248760.1 | 7 | 1364 | |||
XM_006715426.3 | XP_006715489.1 | 9 | 3635 | |||
XM_011535648.2 | XP_011533950.1 | 9 | 3292 | |||
XM_017010626.1 | XP_016866115.1 | 8 | 1627 | |||
GenBank mRNA | AF091622.1 | - | 8 | 3076 | ||
AK298580.1 | - | 7 | 1348 | |||
AK307869.1 | - | 7 | 2975 | |||
BC113650.1 | - | 8 | 3076 | 8322702 | ||
BC113652.1 | - | 8 | 3076 | 8322704 | ||
BX648268.1 | - | 10 | 3302 | |||
D87685.2 | - | 8 | 3064 |