Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX2 SIRNA Assays ›
Gene Symbol:
RBFOX2
Gene Name:
RNA binding protein, fox-1 homolog 2
Gene Aliases:
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location:
Chr.22: 35738736 - 36028892 on Build GRCh38
UniGene ID:
Hs.282998
Species:
Human
siRNA ID
HSS177481
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031695.2
NP_001026865.1
11
1329
NM_001082576.1
NP_001076045.1
11
1317
NM_001082577.1
NP_001076046.1
10
1289
NM_001082578.1
NP_001076047.1
12
1289
NM_001082579.1
NP_001076048.1
12
1286
NM_014309.2
NP_055124.1
10
1280
XM_005261428.2
XP_005261485.1
13
1664
XM_005261429.2
XP_005261486.1
13
1661
XM_005261430.2
XP_005261487.1
12
1635
XM_005261431.2
XP_005261488.1
12
1632
XM_005261432.2
XP_005261489.1
12
1629
XM_005261433.2
XP_005261490.1
11
1592
XM_005261435.1
XP_005261492.1
12
1365
XM_005261437.1
XP_005261494.1
11
1336
XM_006724185.2
XP_006724248.1
13
1676
XM_006724186.2
XP_006724249.1
13
1673
XM_006724187.2
XP_006724250.1
12
1647
XM_006724188.2
XP_006724251.1
11
1604
XM_006724189.3
XP_006724252.1
13
1369
XM_006724190.1
XP_006724253.1
13
1063
XM_006724191.1
XP_006724254.1
12
1368
XM_006724192.2
XP_006724255.1
12
1198
XM_006724193.3
XP_006724256.1
13
1687
XM_006724194.2
XP_006724257.1
12
1140
XM_017028686.1
XP_016884175.1
13
1055
XM_017028687.1
XP_016884176.1
13
1236
XM_017028688.1
XP_016884177.1
11
1589
XM_017028689.1
XP_016884178.1
13
1129
XM_017028690.1
XP_016884179.1
12
1118
XM_017028691.1
XP_016884180.1
12
1115
XM_017028692.1
XP_016884181.1
12
1366
XM_017028693.1
XP_016884182.1
12
1356
XM_017028694.1
XP_016884183.1
12
1353
XM_017028695.1
XP_016884184.1
11
1041
XM_017028696.1
XP_016884185.1
11
1324
XM_017028697.1
XP_016884186.1
13
2145
XM_017028698.1
XP_016884187.1
10
1281
GenBank mRNA
AB649123.1
-
12
996
AF229058.1
-
11
1134
AK055213.1
-
12
1250
AK091695.1
-
1
2100
AK291460.1
-
11
1213
AK314632.1
-
11
1148
AL009266.1
-
10
1085
AM419009.1
-
12
1008
AY072786.1
-
12
1099
AY960684.1
-
10
924
BC013115.1
-
10
1087
4346741
BC025281.1
-
11
1152
4555865
CR456559.1
-
11
1018
DQ778625.1
-
12
1177
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
fibroblast growth factor receptor signaling pathway
regulation of definitive erythrocyte differentiation
RNA metabolic process
radial glia guided migration of Purkinje cell
intracellular estrogen receptor signaling pathway
regulation of cell proliferation
negative regulation of transcription, DNA-templated
dendrite morphogenesis
neuromuscular process controlling balance
nucleotide binding
transcription corepressor activity
RNA binding
mRNA binding
protein binding
transcription factor binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-hjwgp:80/10.239.166.163:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0