Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005253.3 | NP_005244.1 | 4 | 1690 | ||
XM_006711976.2 | XP_006712039.1 | 4 | 1894 | |||
XM_006711977.3 | XP_006712040.1 | 4 | 2747 | |||
XM_017003737.1 | XP_016859226.1 | 4 | 2403 | |||
GenBank mRNA | AK055579.1 | - | 4 | 1744 | ||
AK302622.1 | - | 4 | 844 | |||
BC022791.1 | - | 4 | 1114 | 4637727 | ||
BX647822.1 | - | 4 | 866 | |||
CR542262.1 | - | 4 | 881 | |||
X16706.1 | - | 4 | 884 |