Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 8 | 1743 | ||
XM_005264237.3 | XP_005264294.1 | 9 | 2016 | |||
XM_005264238.3 | XP_005264295.1 | 8 | 992 | |||
XM_017003741.1 | XP_016859230.1 | 10 | 2021 | |||
XM_017003742.1 | XP_016859231.1 | 9 | 1060 | |||
XM_017003743.1 | XP_016859232.1 | 6 | 742 | |||
XM_017003744.1 | XP_016859233.1 | 5 | 490 | |||
GenBank mRNA | AB015982.2 | - | 8 | 1743 | ||
AK027585.1 | - | 5 | 552 | |||
BC030706.2 | - | 8 | 1688 | 5266769 |