Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 18 | 3186 | ||
XM_005264237.3 | XP_005264294.1 | 19 | 3459 | |||
XM_005264238.3 | XP_005264295.1 | 18 | 2435 | |||
XM_017003741.1 | XP_016859230.1 | 20 | 3464 | |||
XM_017003742.1 | XP_016859231.1 | 19 | 2503 | |||
XM_017003743.1 | XP_016859232.1 | 16 | 2185 | |||
XM_017003744.1 | XP_016859233.1 | 15 | 1933 | |||
GenBank mRNA | AB015982.2 | - | 18 | 3186 | ||
BI262400.1 | - | 224 |