Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L13 SIRNA Assays ›
Gene Symbol:
BCL2L13
Gene Name:
BCL2 like 13
Gene Aliases:
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location:
Chr.22: 17628855 - 17730855 on Build GRCh38
UniGene ID:
Hs.631672
Species:
Human
siRNA ID
HSS177581
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270726.1
NP_001257655.1
6
924
NM_001270727.1
NP_001257656.1
5
780
NM_001270728.1
NP_001257657.1
4
760
NM_001270729.1
NP_001257658.1
6
938
NM_001270730.1
NP_001257659.1
5
767
NM_001270731.1
NP_001257660.1
6
1025
NM_001270732.1
NP_001257661.1
5
868
NM_001270733.1
NP_001257662.1
2
310
NM_001270734.1
NP_001257663.1
5
881
NM_001270735.1
NP_001257664.1
4
724
NM_015367.3
NP_056182.2
7
1095
NR_073068.1
5
693
NR_073069.1
5
676
XM_011546119.1
XP_011544421.1
7
1458
XM_011546120.1
XP_011544422.1
7
1460
XM_011546121.2
XP_011544423.1
8
1443
XM_011546123.1
XP_011544425.1
6
823
XM_017028725.1
XP_016884214.1
6
1314
XM_017028726.1
XP_016884215.1
9
1744
XM_017028727.1
XP_016884216.1
7
893
XM_017028728.1
XP_016884217.1
7
1472
XM_017028729.1
XP_016884218.1
7
967
XM_017028730.1
XP_016884219.1
5
681
XM_017028731.1
XP_016884220.1
6
1301
XM_017028732.1
XP_016884221.1
4
544
XM_017028734.1
XP_016884223.1
6
1402
GenBank mRNA
AF146568.1
-
4
516
AF183411.1
-
677
AF325209.1
-
6
742
AF539453.1
-
5
626
AK056248.1
-
6
781
AK291449.1
-
5
688
AK294286.1
-
4
587
AK301394.1
-
2
224
AK314489.1
-
7
922
AL133029.1
-
2
197
AL831982.2
-
5
655
BC003032.1
-
3
355
BC007658.2
-
7
854
3343994
BX407004.2
-
6
677
CN420914.1
-
5
675
CR456521.1
-
7
998
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0