Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271209.1 | NP_001258138.1 | 2 | 302 | ||
NM_153323.4 | NP_697018.1 | 2 | 305 | |||
GenBank mRNA | AF479699.1 | - | 2 | 136 | ||
AY122473.1 | - | 2 | 139 |