Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002032.2 | NP_002023.2 | 3 | 583 | ||
GenBank mRNA | AB062402.1 | - | 3 | 536 | ||
AF088851.1 | - | 3 | 390 | |||
AK311818.1 | - | 3 | 554 | |||
AY258285.1 | - | 1141 | ||||
BC000857.1 | - | 3 | 542 | 3459353 | ||
BC001399.2 | - | 3 | 511 | 3051458 | ||
BC011359.2 | - | 3 | 533 | 4157296 | ||
BC013724.2 | - | 3 | 536 | 3857790 | ||
BC015156.1 | - | 3 | 406 | 3883694 | ||
BC016009.1 | - | 3 | 558 | 4666545 | ||
BC016857.1 | - | 3 | 502 | 3848510 | ||
BC020300.1 | - | 3 | 519 | |||
BC035441.1 | - | 3 | 542 | |||
BC063514.1 | - | 3 | 525 | 3882345 | ||
BC066341.1 | - | 4 | 460 | 5288688 | ||
BC066961.1 | - | 3 | 556 | 4796312 | ||
BC070494.1 | - | 4 | 497 | |||
BC073750.1 | - | 3 | 540 | 6256153 | ||
BC104643.1 | - | 3 | 558 | 4043391 | ||
BC105802.1 | - | 4 | 535 | |||
BM971420.1 | - | 2 | 659 | |||
L20941.1 | - | 3 | 556 | |||
M11146.1 | - | 3 | 425 | |||
M12937.1 | - | 3 | 374 | |||
M97164.1 | - | 3 | 477 | |||
X00318.1 | - | 3 | 439 |
Gene Symbol: FTH1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002201.1 | 1 | 589 | |||
GenBank mRNA | BC073777.1 | - | 1 | 600 |
Gene Symbol: FTH1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | J04755.1 | - | 1 | 1470 |