Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002039.3 | NP_002030.2 | 3 | 756 | ||
NM_207123.2 | NP_997006.1 | 3 | 756 | |||
XM_006714167.3 | XP_006714230.1 | 3 | 1573 | |||
XM_006714168.3 | XP_006714231.1 | 3 | 1237 | |||
XM_017007966.1 | XP_016863455.1 | 4 | 7400 | |||
XM_017007967.1 | XP_016863456.1 | 3 | 485 | |||
XM_017007968.1 | XP_016863457.1 | 4 | 7398 | |||
XM_017007969.1 | XP_016863458.1 | 3 | 2649 | |||
XM_017007970.1 | XP_016863459.1 | 3 | 2404 | |||
XM_017007971.1 | XP_016863460.1 | 3 | 2404 | |||
GenBank mRNA | AK289767.1 | - | 3 | 699 | ||
AK295684.1 | - | 3 | 400 | |||
AK314635.1 | - | 3 | 436 | |||
BC064848.1 | - | 3 | 620 | 30386955 | ||
U43885.1 | - | 3 | 518 |