Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MBLAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203397.2 | NP_981942.1 | 2 | 797 | ||
XM_005250250.3 | XP_005250307.1 | 2 | 840 | |||
GenBank mRNA | BC031288.1 | - | 2 | 797 | 5299411 | |
BQ441096.1 | - | 2 | 540 |