Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012337.2 | NP_036469.2 | 5, 6 | 596 | ||
GenBank mRNA | AF094758.1 | - | 5, 6 | 629 | ||
AK225566.1 | - | 5, 6 | 642 | |||
AK225582.1 | - | 5, 6 | 642 | |||
AK292249.1 | - | 5, 6 | 645 | |||
AK308262.1 | - | 5, 6 | 639 | |||
BC027874.1 | - | 5, 6 | 596 | |||
BC089391.1 | - | 5, 6 | 791 | 30387473 |