Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114090.1 | NP_001107562.1 | 10 | 1568 | ||
NM_019850.2 | NP_062824.2 | 12 | 1940 | |||
XM_011510923.2 | XP_011509225.1 | 12 | 1906 | |||
XM_011510924.1 | XP_011509226.1 | 8 | 939 | |||
XM_011510925.1 | XP_011509227.1 | 8 | 943 | |||
GenBank mRNA | AJ238899.1 | - | 9 | 1254 | ||
AK293203.1 | - | 8 | 940 | |||
AK297390.1 | - | 10 | 1568 | |||
AK298315.1 | - | 12 | 1892 | |||
AK300953.1 | - | 13 | 1844 | |||
BC031573.1 | - | 12 | 1927 | 5186132 |