Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012094.4 | NP_036226.1 | 1 | 162 | ||
NM_181651.2 | NP_857634.1 | 1 | 162 | |||
NM_181652.2 | NP_857635.1 | 1 | 162 | |||
XM_005273882.4 | XP_005273939.2 | 1 | 133 | |||
GenBank mRNA | AF110731.1 | - | 1 | 70 | ||
AF124993.1 | - | 1 | 103 | |||
AF197952.1 | - | 1 | 143 | |||
AF231705.1 | - | 1 | 53 | |||
AF242525.1 | - | 1 | 69 | |||
BC110983.1 | - | 1 | 162 | 5166149 | ||
BC113723.1 | - | 1 | 68 | 8322775 | ||
BC113725.1 | - | 1 | 68 | 8322777 | ||
BC143849.1 | - | 1 | 68 | 9052365 | ||
BC171733.1 | - | 1 | 68 | 9052364 | ||
BU598032.1 | - | 1 | 156 | |||
CR457203.1 | - | 1 | 34 |