Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 5 | 1355 | ||
XM_005267502.2 | XP_005267559.2 | 5 | 1517 | |||
XM_011536619.2 | XP_011534921.1 | 5 | 953 | |||
XM_011536621.2 | XP_011534923.1 | 5 | 953 | |||
XM_017021148.1 | XP_016876637.1 | 4 | 847 | |||
XM_017021149.1 | XP_016876638.1 | 4 | 847 | |||
XM_017021150.1 | XP_016876639.1 | 4 | 847 | |||
XM_017021151.1 | XP_016876640.1 | 5 | 1549 | |||
XM_017021152.1 | XP_016876641.1 | 5 | 1549 | |||
XM_017021153.1 | XP_016876642.1 | 5 | 953 | |||
GenBank mRNA | AY254380.1 | - | 5 | 1149 |