Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161581.1 | NP_001155053.1 | 10 | 1412 | ||
NM_015426.4 | NP_056241.3 | 10 | 1341 | |||
XM_011533560.1 | XP_011531862.1 | 9 | 964 | |||
XM_011533563.1 | XP_011531865.1 | 8 | 830 | |||
GenBank mRNA | AK075289.1 | - | 10 | 1056 | ||
AK315688.1 | - | 10 | 1063 | |||
AL117629.1 | - | 5 | 380 | |||
BC007417.2 | - | 9 | 991 | |||
BC119692.2 | - | 10 | 1051 | 40116721 | ||
BC119693.2 | - | 10 | 1127 |