Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161580.1 | NP_001155052.1 | 10 | 1381 | ||
NM_001161581.1 | NP_001155053.1 | 11 | 1596 | |||
NM_015426.4 | NP_056241.3 | 11 | 1525 | |||
XM_011533560.1 | XP_011531862.1 | 10 | 1148 | |||
XM_011533561.1 | XP_011531863.1 | 9 | 1004 | |||
XM_017006104.1 | XP_016861593.1 | 10 | 1174 | |||
GenBank mRNA | AK075289.1 | - | 11 | 1240 | ||
AK315688.1 | - | 11 | 1247 | |||
AL117629.1 | - | 6 | 564 | |||
BC007417.2 | - | 10 | 1175 | |||
BC110877.1 | - | 10 | 1381 | 5588109 | ||
BC119692.2 | - | 11 | 1235 | 40116721 | ||
BC119693.2 | - | 11 | 1311 | |||
CA944751.1 | - | 1 | 685 |