Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161580.1 | NP_001155052.1 | 9 | 1213 | ||
NM_001161581.1 | NP_001155053.1 | 9 | 1284 | |||
NM_015426.4 | NP_056241.3 | 9 | 1213 | |||
XM_017006104.1 | XP_016861593.1 | 9 | 1006 | |||
GenBank mRNA | AK075289.1 | - | 9 | 928 | ||
AK315688.1 | - | 9 | 935 | |||
AL117629.1 | - | 4 | 252 | |||
BC007417.2 | - | 8 | 863 | |||
BC110877.1 | - | 9 | 1213 | 5588109 | ||
BC119692.2 | - | 9 | 923 | 40116721 | ||
BC119693.2 | - | 9 | 999 |