Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267510.1 | XP_005267567.1 | 7 | 2293 | ||
GenBank mRNA | AF131760.1 | - | 3 | 506 | ||
AF172816.1 | - | 3 | 1174 | |||
AF177336.1 | - | 3 | 620 | |||
AK291030.1 | - | 7 | 1723 | |||
AL080164.1 | - | 3 | 329 | |||
AL834518.1 | - | 7 | 1728 | |||
AY358399.1 | - | 7 | 2123 | |||
BC018816.2 | - | 3 | 688 | |||
BC064901.1 | - | 4 | 1282 | |||
BC080648.1 | - | 3 | 224 | |||
BC088378.1 | - | 3 | 571 | |||
BC113714.1 | - | 7 | 1605 | 8322766 | ||
BC113716.1 | - | 7 | 1605 | 8322768 | ||
BX161500.1 | - | 7 | 2051 | |||
CR457151.1 | - | 2 | 132 |