Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320766.1 | NP_001307695.1 | 22 | 4199 | ||
NM_015565.2 | NP_056380.2 | 23 | 4238 | |||
XM_017028316.1 | XP_016883805.1 | 22 | 3918 | |||
XM_017028317.1 | XP_016883806.1 | 13 | 2513 | |||
GenBank mRNA | AB018257.2 | - | 23 | 4129 | ||
BC140790.1 | - | 23 | 4151 | 9021658 | ||
BC150284.1 | - | 23 | 4129 | 8860138 | ||
CN361594.1 | - | 3 | 355 |