Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NAT9 SIRNA Assays ›
Gene Symbol:
NAT9
Gene Name:
N-acetyltransferase 9 (putative)
Gene Aliases:
EBSP, hNATL
Chromosome Location:
Chr.17: 74770529 - 74776425 on Build GRCh38
UniGene ID:
Hs.144058
Species:
Human
siRNA ID
HSS178072
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NAT9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001305077.1
NP_001292006.1
3, 4
360
NM_001305078.1
NP_001292007.1
3, 4
359
NM_001305079.1
NP_001292008.1
3, 4
363
NM_001305080.1
NP_001292009.1
3, 4
363
NM_001305081.1
NP_001292010.1
3, 4
360
NM_001305082.1
NP_001292011.1
3, 4
508
NM_001305083.1
NP_001292012.1
3, 4
360
NM_001305084.1
NP_001292013.1
3, 4
363
NM_001305085.1
NP_001292014.1
3, 4
360
NM_001305086.1
NP_001292015.1
3, 4
360
NM_001305087.1
NP_001292016.1
3, 4
383
NM_001305088.1
NP_001292017.1
2, 3
274
NM_015654.4
NP_056469.2
3, 4
363
NR_130953.1
3, 4
363
NR_130954.1
3, 4
360
XM_005257212.4
XP_005257269.1
2, 3
202
XM_006721813.3
XP_006721876.1
2, 3
650
XM_011524607.2
XP_011522909.1
3, 4
305
XM_017024437.1
XP_016879926.1
3, 4
289
XM_017024438.1
XP_016879927.1
3, 4
294
XM_017024439.1
XP_016879928.1
3, 4
330
XM_017024440.1
XP_016879929.1
2, 3
668
XM_017024441.1
XP_016879930.1
2, 3
662
GenBank mRNA
AK123115.1
-
3, 4
295
AK225602.1
-
3, 4
233
AK308569.1
-
3, 4
266
AK312958.1
-
3, 4
233
AL050269.1
-
3, 4
240
AL537925.3
-
3, 4
440
AY632082.1
-
2, 3
199
BC004195.2
-
3, 4
269
3528894
BC004225.2
-
3, 4
258
3538186
BG702300.1
-
3, 4
296
BI821280.1
-
3, 4
260
BM464121.1
-
3, 4
286
BM909550.1
-
3, 4
262
BP197633.1
-
3, 4
275
BP231776.1
-
3, 4
281
BQ068039.1
-
2, 3
156
BU159218.1
-
3, 4
258
BX333236.2
-
3, 4
273
DA010490.1
-
3, 4
303
DA259097.1
-
3, 4
360
DB171198.1
-
3, 4
277
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
N-terminal protein amino acid acetylation
protein binding
N-acetyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0