Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 14, 15 | 2002 | ||
XM_011515260.1 | XP_011513562.1 | 14, 15 | 2002 | |||
XM_011515262.2 | XP_011513564.1 | 14, 15 | 2002 | |||
XM_017011959.1 | XP_016867448.1 | 14, 15 | 2002 | |||
XM_017011960.1 | XP_016867449.1 | 14, 15 | 2002 | |||
GenBank mRNA | BF346658.1 | - | 5, 6 | 537 | ||
BX324321.2 | - | 4, 5 | 483 |