Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278055.1 | NP_001264984.1 | 8 | 4282 | ||
NM_014363.5 | NP_055178.3 | 10 | 4813 | |||
XM_005266338.2 | XP_005266395.1 | 11 | 4840 | |||
XM_011535039.2 | XP_011533341.1 | 10 | 4448 | |||
XM_017020539.1 | XP_016876028.1 | 9 | 4421 | |||
GenBank mRNA | CR749427.1 | - | 3 | 3050 |