Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287214.1 | NP_001274143.1 | 5 | 763 | ||
NM_001287215.1 | NP_001274144.1 | 6 | 613 | |||
NM_001287216.1 | NP_001274145.1 | 2 | 490 | |||
NM_001634.5 | NP_001625.2 | 6 | 850 | |||
NR_109768.1 | 3 | 533 | ||||
GenBank mRNA | AJ420527.1 | - | 3 | 508 | ||
AK125644.1 | - | 2 | 1473 | |||
AK300330.1 | - | 5 | 762 | |||
AK302761.1 | - | 2 | 491 | |||
AK304351.1 | - | 6 | 613 | |||
AK316471.1 | - | 6 | 482 | |||
AL832698.1 | - | 3 | 532 | |||
BC000171.2 | - | 6 | 623 | 2900587 | ||
BX640599.1 | - | 6 | 850 | |||
M21154.1 | - | 6 | 776 |