Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: DNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281428.1 | NP_001268357.1 | 13 | 1393 | ||
NM_012144.3 | NP_036276.1 | 13 | 1381 | |||
XM_006716758.3 | XP_006716821.1 | 12 | 1024 | |||
XM_011517846.1 | XP_011516148.1 | 13 | 1393 | |||
XM_011517847.2 | XP_011516149.1 | 13 | 1393 | |||
XM_011517848.1 | XP_011516150.1 | 13 | 1393 | |||
XM_011517849.1 | XP_011516151.1 | 13 | 1393 | |||
XM_011517850.2 | XP_011516152.1 | 13 | 1393 | |||
XM_017014625.1 | XP_016870114.1 | 13 | 1381 | |||
GenBank mRNA | AF091619.1 | - | 13 | 1298 | ||
AK290696.1 | - | 13 | 1305 | |||
AK302186.1 | - | 10 | 924 | |||
AK303937.1 | - | 10 | 878 | |||
BC030583.2 | - | 13 | 1309 | 4837991 |