Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280796.1 | NP_001267725.1 | 13, 14 | 1733 | ||
NM_001280797.1 | NP_001267726.1 | 12, 13 | 1530 | |||
NM_001280798.1 | NP_001267727.1 | 10, 11 | 1308 | |||
NM_013267.3 | NP_037399.2 | 13, 14 | 1660 | |||
XM_005268797.1 | XP_005268854.1 | 13, 14 | 1287 | |||
XM_017019180.1 | XP_016874669.1 | 12, 13 | 1157 | |||
GenBank mRNA | AF110330.1 | - | 13, 14 | 1415 | ||
AF110331.1 | - | 3, 4 | 347 | |||
AF223944.1 | - | 13, 14 | 1587 | |||
AK000467.1 | - | 13, 14 | 1407 | |||
BC025661.1 | - | 12, 13 | 1245 | |||
HM775422.1 | - | 12, 13 | 1279 | |||
HM775423.1 | - | 11, 12 | 1149 | |||
HM775424.1 | - | 11, 12 | 1212 |