Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014442.2 | NP_055257.2 | 1 | 436 | ||
XM_011526734.2 | XP_011525036.1 | 1 | 430 | |||
XM_011526735.2 | XP_011525037.1 | 1 | 432 | |||
GenBank mRNA | AF195092.1 | - | 1 | 426 | ||
AF223403.1 | - | 1 | 429 | |||
AF310234.1 | - | 1 | 377 | |||
AK290149.1 | - | 1 | 429 | |||
AK290412.1 | - | 1 | 408 | |||
AK295589.1 | - | 1 | 434 | |||
BC053319.1 | - | 1 | 425 | 6302572 | ||
DA388103.1 | - | 1 | 436 |