Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMADHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015702.2 | NP_056517.1 | 8 | 1032 | ||
GenBank mRNA | AF060224.1 | - | 7 | 862 | ||
AF131802.1 | - | 8 | 922 | |||
AF161510.1 | - | 8 | 984 | |||
AK310001.1 | - | 8 | 834 | |||
AK313284.1 | - | 8 | 931 | |||
BC000932.2 | - | 8 | 981 | 3446713 | ||
BC010894.1 | - | 8 | 955 | 4246499 | ||
BC022859.1 | - | 8 | 978 | 3826071 | ||
BC023995.1 | - | 8 | 946 | 4103877 |