Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
siRNA ID
HSS178418
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
4
468
NM_001256235.1
NP_001243164.1
5
409
NM_001256236.1
NP_001243165.1
5
651
NM_001256237.1
NP_001243166.1
4
468
NM_001256238.1
NP_001243167.1
4
382
NM_001256239.1
NP_001243168.1
3
398
NM_001256240.1
NP_001243169.1
3
398
NM_001283038.1
NP_001269967.1
4
468
NM_001283039.1
NP_001269968.1
4
421
NM_014489.3
NP_055304.1
4
581
NR_027016.2
2
223
NR_027017.2
5
531
NR_027018.2
5
420
NR_045923.1
3
293
NR_045927.1
3
293
NR_045929.1
3
351
NR_104270.1
3
293
NR_104271.1
5
553
NR_104272.1
2
223
XM_006718181.3
XP_006718244.1
3
1059
XM_006718183.3
XP_006718246.1
5
476
XM_006718185.2
XP_006718248.1
4
368
XM_006718186.2
XP_006718249.1
5
528
XM_006718190.3
XP_006718253.1
4
684
XM_006718191.3
XP_006718254.1
4
684
XM_006718192.1
XP_006718255.1
6
602
XM_006718193.2
XP_006718256.1
5
409
XM_011519990.2
XP_011518292.2
5
935
XM_011519991.2
XP_011518293.2
5
938
XM_011519992.1
XP_011518294.1
6
707
XM_011519993.2
XP_011518295.2
5
954
XM_011519994.2
XP_011518296.2
5
954
XM_011519995.2
XP_011518297.1
5
858
XM_011519996.1
XP_011518298.1
6
659
XM_011519997.1
XP_011518299.1
6
1000
XM_011519998.2
XP_011518300.1
6
678
XM_011519999.1
XP_011518301.1
6
637
XM_011520000.1
XP_011518302.1
5
551
XM_011520001.1
XP_011518303.1
4
343
XM_011520002.1
XP_011518304.1
4
343
XM_011520003.2
XP_011518305.1
5
867
XM_011520004.2
XP_011518306.1
5
542
XM_011520005.2
XP_011518307.1
4
439
XM_011520006.2
XP_011518308.1
5
823
XM_011520007.2
XP_011518309.1
6
592
XM_017017556.1
XP_016873045.1
4
343
XM_017017557.1
XP_016873046.1
4
491
XM_017017558.1
XP_016873047.1
4
665
XM_017017559.1
XP_016873048.1
5
528
XM_017017560.1
XP_016873049.1
5
528
GenBank mRNA
AF159615.1
-
4
583
AK292181.1
-
5
545
AK295202.1
-
5
420
AK301991.1
-
5
501
AK307359.1
-
3
210
AK308525.1
-
3
204
AL049261.1
-
2
833
AL096753.1
-
5
409
AY373030.1
-
2
272
BC009930.2
-
4
355
3506614
BG707484.1
-
3
187
BI460657.1
-
6
664
BI759334.1
-
3
291
BI768071.1
-
3
355
BI771208.1
-
2
184
BI772237.1
-
4
361
BI911028.1
-
4
497
BI911526.1
-
5
442
BX340489.2
-
3
335
BX356273.2
-
4
375
BX379511.2
-
4
350
BX448155.2
-
4
327
CB988453.1
-
2
250
CD675113.1
-
3
186
CX872317.1
-
2
151
DA287291.1
-
4
468
KU178608.1
-
2
272
KU178609.1
-
3
357
KU178610.1
-
3
357
KU178611.1
-
2
272
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-chv4l:80/10.239.160.128:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0