Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
siRNA ID
HSS178419
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
5
672
NM_001256235.1
NP_001243164.1
6
613
NM_001256236.1
NP_001243165.1
6
855
NM_001256237.1
NP_001243166.1
5
672
NM_001256238.1
NP_001243167.1
5
586
NM_001256239.1
NP_001243168.1
4
602
NM_001256240.1
NP_001243169.1
4
602
NM_001283038.1
NP_001269967.1
5
672
NM_001283039.1
NP_001269968.1
5
625
NM_001283040.1
NP_001269969.1
3
244
NM_014489.3
NP_055304.1
5
785
NR_027016.2
3
427
NR_027017.2
6
735
NR_027018.2
6
624
NR_045923.1
4
497
NR_045925.1
4
419
NR_045926.1
4
419
NR_045927.1
4
497
NR_045929.1
4
555
NR_104270.1
4
497
NR_104271.1
6
757
NR_104272.1
3
427
XM_006718181.3
XP_006718244.1
4
1263
XM_006718183.3
XP_006718246.1
6
680
XM_006718185.2
XP_006718248.1
5
572
XM_006718186.2
XP_006718249.1
6
732
XM_006718190.3
XP_006718253.1
5
888
XM_006718191.3
XP_006718254.1
5
888
XM_006718192.1
XP_006718255.1
7
806
XM_006718193.2
XP_006718256.1
6
613
XM_011519990.2
XP_011518292.2
6
1139
XM_011519991.2
XP_011518293.2
6
1142
XM_011519992.1
XP_011518294.1
7
911
XM_011519993.2
XP_011518295.2
6
1158
XM_011519994.2
XP_011518296.2
6
1158
XM_011519995.2
XP_011518297.1
6
1062
XM_011519996.1
XP_011518298.1
7
863
XM_011519997.1
XP_011518299.1
7
1204
XM_011519998.2
XP_011518300.1
7
882
XM_011519999.1
XP_011518301.1
7
841
XM_011520000.1
XP_011518302.1
6
755
XM_011520001.1
XP_011518303.1
5
547
XM_011520002.1
XP_011518304.1
5
547
XM_011520003.2
XP_011518305.1
6
1071
XM_011520004.2
XP_011518306.1
6
746
XM_011520005.2
XP_011518307.1
5
643
XM_011520006.2
XP_011518308.1
6
1027
XM_011520007.2
XP_011518309.1
7
796
XM_017017556.1
XP_016873045.1
5
547
XM_017017557.1
XP_016873046.1
5
695
XM_017017558.1
XP_016873047.1
5
869
XM_017017559.1
XP_016873048.1
6
732
XM_017017560.1
XP_016873049.1
6
732
GenBank mRNA
AF159615.1
-
5
787
AK292181.1
-
6
749
AK295202.1
-
6
624
AK301991.1
-
6
705
AK307359.1
-
4
414
AK308525.1
-
3
542
AL049261.1
-
2
1171
AL096753.1
-
6
613
AL558360.3
-
4
326
AY373030.1
-
3
476
BC009930.2
-
5
559
3506614
BG468212.1
-
4
308
BG707484.1
-
4
391
BI759334.1
-
4
494
BI768071.1
-
4
559
BI771208.1
-
3
388
BI772237.1
-
5
565
BI911526.1
-
6
646
BX340489.2
-
4
539
BX356273.2
-
5
579
BX379511.2
-
5
554
BX448155.2
-
5
531
CB988453.1
-
3
454
CD675009.1
-
2
138
CD675113.1
-
4
390
CX872317.1
-
3
355
HY171112.1
-
3
165
KU178608.1
-
3
476
KU178609.1
-
4
561
KU178610.1
-
4
561
KU178611.1
-
3
476
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-cq5mm:80/10.239.166.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0